5-Bromo-2-methylnicotinic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR959451-5G
APOSOR959451-5GEA
171
EUR
APOSOR959451-5G
5-Bromo-2-methylnicotinic acid 95%
5-Bromo-2-methylnicotinic acid
Formel:
C₇H₆BrNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11557230 CAS-Nummer: 351003-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...