2-Chloro-5-methylnicotinaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-methylpyridine-3-carbaldehyde
OR959452-1G
OR959452-250MG
APOSOR959452-1GEA
114
EUR
APOSOR959452-1G
APOSOR959452-250MG
2-Chloro-5-methylnicotinaldehyde 98%
2-Chloro-5-methylnicotinaldehyde
Formel:
C₇H₆ClNO Molecular Weight: 155.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375370 CAS-Nummer: 92444-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...