2-Bromo-5-chloro-4-methylpyridine 98%

Lieferant: Apollo Scientific
Achtung

OR959457-1G OR959457-25G OR959457-5G
APOSOR959457-1GEA 86.1 EUR
APOSOR959457-1G APOSOR959457-25G APOSOR959457-5G
2-Bromo-5-chloro-4-methylpyridine 98%
2-Bromo-5-chloro-4-methylpyridine
Formel: C₆H₅BrClN
Lagertemperatur: Kühlschrank
MDL: MFCD11846275
CAS-Nummer: 885267-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR