2-Amino-5-bromoisonicotinonitrile 95%

Lieferant: Apollo Scientific

OR959472-1G
APOSOR959472-1GEA 231 EUR
APOSOR959472-1G
2-Amino-5-bromoisonicotinonitrile 95%
2-Amino-5-bromoisonicotinonitrile
Formel: C₆H₄BrN₃
Molecular Weight: 198.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907918
CAS-Nummer: 944401-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR