3-Chloro-6-methoxy-4-methylpyridazine 95%
Lieferant: Apollo Scientific
OR959479-1G
APOSOR959479-1GEA
806
EUR
APOSOR959479-1G
3-Chloro-6-methoxy-4-methylpyridazine 95%
3-Chloro-6-methoxy-4-methylpyridazine
Formel:
C₆H₇ClN₂O Molecular Weight: 158,586 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234198 CAS-Nummer: 89466-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...