5-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-[1,2,4]triazolo[4,3-a]pyridine

OR959510-1G OR959510-250MG
APOSOR959510-1GEA 267 EUR
APOSOR959510-1G APOSOR959510-250MG
5-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%
5-Bromo[1,2,4]triazolo[4,3-a]pyridine
Formel: C₆H₄BrN₃
Molecular Weight: 198.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11043995
CAS-Nummer: 1172085-67-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR