6-Cyanoquinoxaline 95%
Lieferant: Apollo Scientific
OR959535-1G
APOSOR959535-1GEA
115
EUR
APOSOR959535-1G
6-Cyanoquinoxaline 95%
6-Cyanoquinoxaline
Formel:
C₉H₅N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669502 CAS-Nummer: 23088-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...