3-Methoxypyridine-2-carboxamide 95%

Lieferant: Apollo Scientific

OR959569-1G
APOSOR959569-1GEA 632 EUR
APOSOR959569-1G
3-Methoxypyridine-2-carboxamide 95%
3-Methoxypyridine-2-carboxamide
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976464
CAS-Nummer: 24059-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR