(3-Bromo-4-pyridyl)methanol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-4-pyridinemethanol, 3-Bromo-4-(hydroxymethyl)pyridine

OR959577-5G
APOSOR959577-5GEA 108 EUR
APOSOR959577-5G
(3-Bromo-4-pyridyl)methanol 98%
(3-Bromo-4-pyridyl)methanol
Formel: C₆H₆BrNO
Molecular Weight: 188.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847285
CAS-Nummer: 146679-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR