(2-methylpyridin-3-yl)methanol 95%

Lieferant: Apollo Scientific

OR959578-1G
APOSOR959578-1GEA 59.6 EUR
APOSOR959578-1G
(2-methylpyridin-3-yl)methanol 95%
(2-methylpyridin-3-yl)methanol
Formel: C₇H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00030820
CAS-Nummer: 56826-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR