3-Indolylessigsäurehydrazid

Lieferant: Apollo Scientific
Achtung

Synonyme: Indol-3-ylessigsäurehydrazid, Indol-3-essigsäurehydrazid

OR9596-5G
APOSOR9596-5GEA 59.6 EUR
APOSOR9596-5G
3-Indolylessigsäurehydrazid
3-Indolylessigsäurehydrazid
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189.22 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005634
CAS-Nummer: 5448-47-5
EINECS: 226-672-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR