5-bromo-4-methoxypyrimidine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-4-methoxypyrimidine

OR959616-1G OR959616-250MG
APOSOR959616-1GEA 157 EUR
APOSOR959616-1G APOSOR959616-250MG
5-bromo-4-methoxypyrimidine 95%
5-bromo-4-methoxypyrimidine
Formel: C₅H₅BrN₂O
Molecular Weight: 189.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234066
CAS-Nummer: 4319-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR