6-Chloro-2-methoxyquinoline 98%

Lieferant: Apollo Scientific

OR959637-1G
APOSOR959637-1GEA 0 EUR
APOSOR959637-1G
6-Chloro-2-methoxyquinoline 98%
6-Chloro-2-methoxyquinoline
Formel: C₁₀H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD22418817
CAS-Nummer: 1820686-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR