2-methyl-1,5-naphthyridine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-1,5-naphthyridine
OR959646-1G
OR959646-250MG
OR959646-5G
OR959646-10G
APOSOR959646-1GEA
102
EUR
APOSOR959646-1G
APOSOR959646-250MG
APOSOR959646-5G
APOSOR959646-10G
2-methyl-1,5-naphthyridine 95%
2-methyl-1,5-naphthyridine
Formel:
C₉H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437992 CAS-Nummer: 7675-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...