2-methyl-1,5-naphthyridine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-1,5-naphthyridine

OR959646-1G OR959646-250MG OR959646-5G OR959646-10G
APOSOR959646-1GEA 102 EUR
APOSOR959646-1G APOSOR959646-250MG APOSOR959646-5G APOSOR959646-10G
2-methyl-1,5-naphthyridine 95%
2-methyl-1,5-naphthyridine
Formel: C₉H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07437992
CAS-Nummer: 7675-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR