3-Bromoquinoline-6-carboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR959655-250MG
APOSOR959655-250MGEA
128
EUR
APOSOR959655-250MG
3-Bromoquinoline-6-carboxylic acid 95+%
3-Bromoquinoline-6-carboxylic acid
Formel:
C₁₀H₆BrNO₂ Molecular Weight: 252,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18086942 CAS-Nummer: 205114-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...