2,5-Dimethoxypyridin-3-amine 95%

Lieferant: Apollo Scientific

OR959686-1G
APOSOR959686-1GEA 641 EUR
APOSOR959686-1G
2,5-Dimethoxypyridin-3-amine 95%
2,5-Dimethoxypyridin-3-amine
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038237
CAS-Nummer: 1211595-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR