5-Bromo-4-ethyl-3,4-dihydropyrimidine 97%
Lieferant: Apollo Scientific
OR959712-1G
APOSOR959712-1GEA
451
EUR
APOSOR959712-1G
5-Bromo-4-ethyl-3,4-dihydropyrimidine 97%
5-Bromo-4-ethyl-3,4-dihydropyrimidine
Formel:
C₆H₉BrN₂ Molecular Weight: 189,053 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20681885 CAS-Nummer: 224429-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...