N-(3-Formylpyridin-2-yl)acetamide 97%

Lieferant: Apollo Scientific

OR959719-1G
APOSOR959719-1GEA 632 EUR
APOSOR959719-1G
N-(3-Formylpyridin-2-yl)acetamide 97%
N-(3-Formylpyridin-2-yl)acetamide
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17170608
CAS-Nummer: 54856-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR