5-Amino-3-chloropicoline 98%

Lieferant: Apollo Scientific

OR959728-1G OR959728-5G
APOSOR959728-1GEA 270 EUR
APOSOR959728-1G APOSOR959728-5G
5-Amino-3-chloropicoline 98%
5-Amino-3-chloropicoline
Formel: C₆H₇ClN₂
Molecular Weight: 142.586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040270
CAS-Nummer: 896161-13-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR