Dimethyl[(E)-2-(3-nitropyridin-2-yl)ethenyl]amine 98%
Lieferant: Apollo Scientific
OR959742-1G
APOSOR959742-1GEA
364
EUR
APOSOR959742-1G
Dimethyl[(E)-2-(3-nitropyridin-2-yl)ethenyl]amine 98%
Dimethyl[(E)-2-(3-nitropyridin-2-yl)ethenyl]amine
Formel:
C₉H₁₁N₃O₂ Molecular Weight: 193,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054129 CAS-Nummer: 343569-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...