Quinazolin-2-ol 98%
Lieferant: Apollo Scientific
OR959744-1G
APOSOR959744-1GEA
632
EUR
APOSOR959744-1G
Quinazolin-2-ol 98%
Quinazolin-2-ol
Formel:
C₈H₆N₂O Molecular Weight: 146,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00972003 CAS-Nummer: 7471-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...