N,N,3-Trimethylpyridine-2-carboxamide 95%

Lieferant: Apollo Scientific

OR959756-1G
APOSOR959756-1GEA 299 EUR
APOSOR959756-1G
N,N,3-Trimethylpyridine-2-carboxamide 95%
N,N,3-Trimethylpyridine-2-carboxamide
Formel: C₉H₁₂N₂O
Molecular Weight: 164.204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20366305
CAS-Nummer: 1341499-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR