2-Ethoxypyridine-3-carboxamide 95%
Lieferant: Apollo Scientific
OR959774-1G
APOSOR959774-1GEA
574
EUR
APOSOR959774-1G
2-Ethoxypyridine-3-carboxamide 95%
2-Ethoxypyridine-3-carboxamide
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18109033 CAS-Nummer: 119646-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...