1-(5-hydroxypyridin-2-yl)ethanone 95%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Hydroxypyridin-2-yl)ethanone

OR959785-1G
APOSOR959785-1GEA 128 EUR
APOSOR959785-1G
1-(5-hydroxypyridin-2-yl)ethanone 95%
1-(5-hydroxypyridin-2-yl)ethanone
Formel: C₇H₇NO₂
Molecular Weight: 137.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697710
CAS-Nummer: 67310-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR