Quinoline-4-carboxamide 98%

Lieferant: Apollo Scientific

OR959786-1G
APOSOR959786-1GEA 250 EUR
APOSOR959786-1G
Quinoline-4-carboxamide 98%
Quinoline-4-carboxamide
Formel: C₁₀H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00672908
CAS-Nummer: 4363-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR