N-Methylpyrimidin-4-amine 95%

Lieferant: Apollo Scientific

OR959792-1G
APOSOR959792-1GEA 270 EUR
APOSOR959792-1G
N-Methylpyrimidin-4-amine 95%
N-Methylpyrimidin-4-amine
Formel: C₅H₇N₃
Molecular Weight: 109,129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697837
CAS-Nummer: 22632-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR