2-Methoxy-4-phenylpyridine 96%

Lieferant: Apollo Scientific

OR959817-1G
APOSOR959817-1GEA 574 EUR
APOSOR959817-1G
2-Methoxy-4-phenylpyridine 96%
2-Methoxy-4-phenylpyridine
Formel: C₁₂H₁₁NO
Molecular Weight: 185,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044372
CAS-Nummer: 53698-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR