2,5-Dimethyl-3-nitropyridine 98%

Lieferant: Apollo Scientific

OR959822-1G
APOSOR959822-1GEA 364 EUR
APOSOR959822-1G
2,5-Dimethyl-3-nitropyridine 98%
2,5-Dimethyl-3-nitropyridine
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10574716
CAS-Nummer: 52381-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR