N-Methoxy-N,2-dimethylpropanamide 98%

Lieferant: Apollo Scientific

Synonyme: N-Methoxy-N-methylisobutyramide

OR959864-25G
APOSOR959864-25GEA 140 EUR
APOSOR959864-25G
N-Methoxy-N,2-dimethylpropanamide 98%
N-Methoxy-N,2-dimethylpropanamide
Formel: C₆H₁₃NO₂
Molecular Weight: 131.173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847606
CAS-Nummer: 113778-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR