2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 1-Isobutyl-1H-pyrazole-5-carbaldehyde

OR959892-1G
APOSOR959892-1GEA 364 EUR
APOSOR959892-1G
2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%
2-(2-Methylpropyl)pyrazole-3-carbaldehyde
Formel: C₈H₁₂N₂O
Molecular Weight: 152,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21609707
CAS-Nummer: 1236366-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR