2-(Isopropylamino)benzonitrile 98%

Lieferant: Apollo Scientific

OR959902-1G OR959902-5G
APOSOR959902-1GEA 168 EUR
APOSOR959902-1G APOSOR959902-5G
2-(Isopropylamino)benzonitrile 98%
2-(Isopropylamino)benzonitrile
Formel: C₁₀H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11144672
CAS-Nummer: 147531-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR