2'-Brom-5'-methoxyacetophenon

Lieferant: Apollo Scientific
Achtung

OR9600-1G OR9600-5G
APOSOR9600-1GEA 68.3 EUR
APOSOR9600-1G APOSOR9600-5G
2'-Brom-5'-methoxyacetophenon
2'-Brom-5'-methoxyacetophenon
Formel: C₉H₉BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02683853
CAS-Nummer: 6342-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR