3-(Cyclopropylmethoxy)azetidine hydrochloride 95%

Lieferant: Apollo Scientific

OR960080-1G OR960080-5G
APOSOR960080-1GEA 290 EUR
APOSOR960080-1G APOSOR960080-5G
3-(Cyclopropylmethoxy)azetidine hydrochloride 95%
3-(Cyclopropylmethoxy)azetidine hydrochloride
Formel: C₇H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22205833
CAS-Nummer: 1375069-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR