Methyl 4-methylpiperidine-4-carboxylate 97%

Lieferant: Apollo Scientific

OR960153-1G
APOSOR960153-1GEA 364 EUR
APOSOR960153-1G
Methyl 4-methylpiperidine-4-carboxylate 97%
Methyl 4-methylpiperidine-4-carboxylate
Formel: C₈H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13178892
CAS-Nummer: 892493-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR