7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 97%
Lieferant: Apollo Scientific
OR960206-10G
OR960206-1G
OR960206-5G
APOSOR960206-10GEA
754
EUR
APOSOR960206-10G
APOSOR960206-1G
APOSOR960206-5G
7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 97%
7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
Formel:
C₉H₈ClNO₂ Molecular Weight: 197.618 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26381971 CAS-Nummer: 1506636-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...