{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: [3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methanamine

OR960244-1G
APOSOR960244-1GEA 328 EUR
APOSOR960244-1G
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride 98%
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride
Formel: C₁₀H₁₂ClN₃O
Molecular Weight: 225.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13186189
CAS-Nummer: 890324-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR