3-Formyl-5-methylbenzoic acid 96%

Lieferant: Apollo Scientific

OR960245-1G
APOSOR960245-1GEA 632 EUR
APOSOR960245-1G
3-Formyl-5-methylbenzoic acid 96%
3-Formyl-5-methylbenzoic acid
Formel: C₉H₈O₃
Molecular Weight: 164,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20645804
CAS-Nummer: 4481-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR