4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxycyclohexanecarboxylic acid
OR960249-25G
APOSOR960249-25GEA
94.4
EUR
APOSOR960249-25G
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95+%
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)
Formel:
C₇H₁₂O₃ Molecular Weight: 144,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077974 CAS-Nummer: 17419-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...