2-Ethoxy-3-methylpyridine-5-carboxylic acid 95%

Lieferant: Apollo Scientific

OR960260-1G
APOSOR960260-1GEA 806 EUR
APOSOR960260-1G
2-Ethoxy-3-methylpyridine-5-carboxylic acid 95%
2-Ethoxy-3-methylpyridine-5-carboxylic acid
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21087314
CAS-Nummer: 1355216-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR