Ethyl-4-methylenecyclohexanecarboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-methylenecyclohexanecarboxylate

OR960267-1G
APOSOR960267-1GEA 21.9 EUR
APOSOR960267-1G
Ethyl-4-methylenecyclohexanecarboxylate 96%
Ethyl-4-methylenecyclohexanecarboxylate
Formel: C₁₀H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12498694
CAS-Nummer: 145576-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR