3-Methyl-5-phenyl-1,2,4-oxadiazole 98%

Lieferant: Apollo Scientific

OR960271-1G
APOSOR960271-1GEA 267 EUR
APOSOR960271-1G
3-Methyl-5-phenyl-1,2,4-oxadiazole 98%
3-Methyl-5-phenyl-1,2,4-oxadiazole
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464109
CAS-Nummer: 1199-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR