4-(Isopropylsulphanyl)benzaldehyde 98%

Lieferant: Apollo Scientific

OR960283-1G
APOSOR960283-1GEA 502 EUR
APOSOR960283-1G
4-(Isopropylsulphanyl)benzaldehyde 98%
4-(Isopropylsulphanyl)benzaldehyde
Formel: C₁₀H₁₂OS
Molecular Weight: 180,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18887933
CAS-Nummer: 84264-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR