(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine 96%

Lieferant: Apollo Scientific

Synonyme: 1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-N-methylmethanamine

OR960297-1G
APOSOR960297-1GEA 206 EUR
APOSOR960297-1G
(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine 96%
(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)(methyl)amine
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06260669
CAS-Nummer: 613656-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR