5-Chloro-1-methylindazole 98%

Lieferant: Apollo Scientific

OR960303-1G
APOSOR960303-1GEA 435 EUR
APOSOR960303-1G
5-Chloro-1-methylindazole 98%
5-Chloro-1-methylindazole
Formel: C₈H₇ClN₂
Molecular Weight: 166,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977519
CAS-Nummer: 1379091-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR