N-Methylthian-4-amine hydrochloride 97%

Lieferant: Apollo Scientific

OR960312-1G
APOSOR960312-1GEA 319 EUR
APOSOR960312-1G
N-Methylthian-4-amine hydrochloride 97%
N-Methylthian-4-amine hydrochloride
Formel: C₆H₁₃NS·ClH
Molecular Weight: 167,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13857451
CAS-Nummer: 1188964-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR