Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR960335-1G
APOSOR960335-1GEA
72.5
EUR
APOSOR960335-1G
Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Formel:
C₉H₉N₃O₂S Molecular Weight: 223,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00096361 CAS-Nummer: 56881-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...