2,2-Dimethylpent-4-enenitrile 97%

Lieferant: Apollo Scientific

OR960348-1G
APOSOR960348-1GEA 806 EUR
APOSOR960348-1G
2,2-Dimethylpent-4-enenitrile 97%
2,2-Dimethylpent-4-enenitrile
Formel: C₇H₁₁N
Lagertemperatur: Raumtemperatur
MDL: MFCD20483740
CAS-Nummer: 2978-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR