Fmoc-N-Me-Val-OH 97%

Lieferant: Apollo Scientific

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-valine, Fmoc-N-Me-Val-OH, N-Fmoc-N-methyl-L-valine

OR960386-25G
APOSOR960386-25GEA 108 EUR
APOSOR960386-25G
Fmoc-N-Me-Val-OH 97%
Fmoc-N-Me-Val-OH
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353.418 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153395
CAS-Nummer: 84000-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR