N-Benzyloxycarbonyl-L-norleucin 97%

Lieferant: Apollo Scientific

Synonyme: N-Cbz-L-norleucine, Z-Nle-OH

OR960529-1G OR960529-25G OR960529-5G
APOSOR960529-1GEA 95.7 EUR
APOSOR960529-1G APOSOR960529-25G APOSOR960529-5G
N-Benzyloxycarbonyl-L-norleucin 97%
N-Benzyloxycarbonyl-L-norleucin
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265.309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063170
CAS-Nummer: 39608-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR